Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001313994.1 | Intron | NP_001300923.1 | |||
NM_145004.6 | Intron | NP_659441.3 | |||
XM_006716298.2 | Intron | XP_006716361.1 | |||
XM_006716299.1 | Intron | XP_006716362.1 | |||
XM_011544433.1 | Intron | XP_011542735.1 | |||
XM_011544434.1 | Intron | XP_011542736.1 | |||
XM_017013183.1 | Intron | XP_016868672.1 | |||
XM_017013184.1 | Intron | XP_016868673.1 | |||
XM_017013185.1 | Intron | XP_016868674.1 | |||
XM_017013186.1 | Intron | XP_016868675.1 | |||
XM_017013187.1 | Intron | XP_016868676.1 | |||
XM_017013188.1 | Intron | XP_016868677.1 |