Product Details

SNP ID
rs11094248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72152891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGCATTGGCCAGTCCTAGTATG[A/C]AGTCAAGAGGTGGACTTTTGTTAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLJ44635 PubMed Links

Gene Details

Gene
FLJ44635
Gene Name
TPT1-like protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207422.2 12259 Intron NP_997305.2
Gene
NHSL2
Gene Name
NHS like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013627.2 12259 Intron NP_001013649.2
XM_011530932.1 12259 Intron XP_011529234.1
XM_011530933.1 12259 Intron XP_011529235.1
XM_011530934.2 12259 UTR 3 XP_011529236.1
XM_017029473.1 12259 UTR 3 XP_016884962.1
XM_017029474.1 12259 UTR 3 XP_016884963.1
XM_017029475.1 12259 UTR 3 XP_016884964.1
XM_017029476.1 12259 UTR 3 XP_016884965.1
XM_017029477.1 12259 UTR 3 XP_016884966.1

View Full Product Details