Product Details

SNP ID
rs11080561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12257231 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGGTTGGCTTCTGTCCAATTCCTG[C/T]GTACTGCTTCTTCCTCCCACGGTGG
Phenotype
MIM: 604440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIDEA PubMed Links

Gene Details

Gene
CIDEA
Gene Name
cell death-inducing DFFA-like effector a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001279.3 Intron NP_001270.1
NM_001318383.1 Intron NP_001305312.1

View Full Product Details