Product Details
- SNP ID
-
rs11041531
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7684209 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTGACCATCAAAAAAGTTTGTC[A/G]GCAGAAGTTAGAAGTGGAGGTATGG
- Phenotype
-
MIM: 608342
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYB5R2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75090015] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYB5R2
- Gene Name
- cytochrome b5 reductase 2
There are no transcripts associated with this gene.
- Gene
- OVCH2
- Gene Name
- ovochymase 2 (gene/pseudogene)
View Full Product Details