Product Details

SNP ID
rs6839340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152768811 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATTTAGGAAGAATCCATAGCTT[A/C]TAGTTACAGATGACAAACCTGATAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TIGD4 PubMed Links

Gene Details

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 3118 Intron NP_663772.1
XM_005262807.4 3118 UTR 3 XP_005262864.1

View Full Product Details