Product Details

SNP ID
rs11253132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5366665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACACTTTGATGGTGAAACCAT[C/T]CAGGGTTGCAAAGAACCCTTACATT
Phenotype
MIM: 605901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UCN3 PubMed Links

Gene Details

Gene
UCN3
Gene Name
urocortin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053049.3 Intron NP_444277.2

View Full Product Details