Product Details

SNP ID
rs11671053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23654343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTCTTCACATTTGCAGAAATTC[A/G]CCTTGGTATAATTTCTTTCATGTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 1390 Intron NP_612203.2
XM_017026442.1 1390 Missense Mutation GCG,GTG A121V XP_016881931.1
XM_017026443.1 1390 Missense Mutation GCG,GTG A121V XP_016881932.1
XM_017026444.1 1390 Intron XP_016881933.1

View Full Product Details