Product Details

SNP ID
rs8106553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23658023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGCAAAACCATAGTAAGAGAAAC[A/G]TTTACTCATAAAATCTTGCAGGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 Intron NP_612203.2
XM_017026442.1 Intron XP_016881931.1
XM_017026443.1 Intron XP_016881932.1
XM_017026444.1 Intron XP_016881933.1

View Full Product Details