Product Details

SNP ID
rs12069472
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:39563779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAATACAATTAAAGCCCATCAGT[C/T]TGGGCAAAGACCCGTCAAATCCCAT
Phenotype
MIM: 603407
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929516 PubMed Links

Gene Details

Gene
LOC101929516
Gene Name
uncharacterized LOC101929516
There are no transcripts associated with this gene.

Gene
PABPC4
Gene Name
poly(A) binding protein cytoplasmic 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135653.1 Intron NP_001129125.1
NM_001135654.1 Intron NP_001129126.1
NM_003819.3 Intron NP_003810.1
Gene
PPIEL
Gene Name
peptidylprolyl isomerase E like pseudogene
There are no transcripts associated with this gene.

Gene
SNORA55
Gene Name
small nucleolar RNA, H/ACA box 55
There are no transcripts associated with this gene.

View Full Product Details