Product Details
- SNP ID
-
rs11084798
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35047644 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTCCCGGAACCTAGAAAGGTGCCA[A/G]GCACACAGCGGATGTTCAGTAGGAT
- Phenotype
-
MIM: 142440
MIM: 600235
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HPN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10690630,rs11392434] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HPN
- Gene Name
- hepsin
- Gene
- SCN1B
- Gene Name
- sodium voltage-gated channel beta subunit 1
There are no transcripts associated with this gene.
View Full Product Details