Product Details

SNP ID
rs12836541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23851604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTTGTAACTTAAAAGAAAGCC[C/T]TAGACTGTACCAAGATGACAATATT
Phenotype
MIM: 300753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOO PubMed Links

Gene Details

Gene
APOO
Gene Name
apolipoprotein O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024122.4 Intron NP_077027.1
XM_011545584.2 Intron XP_011543886.1
XM_011545585.2 Intron XP_011543887.1
XM_011545586.2 Intron XP_011543888.1
XM_011545587.2 Intron XP_011543889.1
XM_017029837.1 Intron XP_016885326.1

View Full Product Details