Product Details
- SNP ID
-
rs12427232
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49384313 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAAATATGATTTCTTTTCCTTTTA[A/G]GCAGATGATGCTTTTTCTTCTCTCT
- Phenotype
-
MIM: 611667
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100335030
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115977383] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100335030
- Gene Name
- FGFR1 oncogene partner 2 pseudogene
There are no transcripts associated with this gene.
- Gene
- SPATS2
- Gene Name
- spermatogenesis associated serine rich 2
View Full Product Details