Product Details

SNP ID
rs11137165
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137641224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTGGTTCCTCAGGATGCCTCTT[C/T]CTACTGGCTTTATTCTGCATTTGAC
Phenotype
MIM: 607001
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EHMT1 PubMed Links

Gene Details

Gene
EHMT1
Gene Name
euchromatic histone lysine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145527.1 Intron NP_001138999.1
NM_024757.4 Intron NP_079033.4
XM_005266105.4 Intron XP_005266162.1
XM_005266110.1 Intron XP_005266167.1
XM_006717288.2 Intron XP_006717351.1
XM_011519021.2 Intron XP_011517323.1
XM_011519022.2 Intron XP_011517324.1
XM_011519023.2 Intron XP_011517325.1
XM_011519024.2 Intron XP_011517326.1
XM_011519025.1 Intron XP_011517327.1
XM_011519026.2 Intron XP_011517328.1
XM_011519027.2 Intron XP_011517329.1
XM_011519028.2 Intron XP_011517330.1
XM_011519029.2 Intron XP_011517331.1
XM_011519030.2 Intron XP_011517332.1
XM_011519031.1 Intron XP_011517333.1
XM_011519033.2 Intron XP_011517335.1
XM_017015134.1 Intron XP_016870623.1
XM_017015135.1 Intron XP_016870624.1
XM_017015136.1 Intron XP_016870625.1
XM_017015137.1 Intron XP_016870626.1
XM_017015138.1 Intron XP_016870627.1

View Full Product Details