Product Details

SNP ID
rs10846820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:125068128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTGGCCCATGGGCCAGGTCTCAT[C/G]TCCAGACACACTGTTGAGTTGCCTG
Phenotype
MIM: 614364
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AACS PubMed Links

Gene Details

Gene
AACS
Gene Name
acetoacetyl-CoA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319839.1 Intron NP_001306768.1
NM_001319840.1 Intron NP_001306769.1
NM_023928.4 Intron NP_076417.2
XM_011538692.2 Intron XP_011536994.1

View Full Product Details