Product Details

SNP ID
rs11209376
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:41032676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGACTTCAGGGGGACATGGAGAG[A/G]CCAGTTAAGAAGGAGGCTACTGGCC
Phenotype
MIM: 616396
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCMH1 PubMed Links

Gene Details

Gene
SCMH1
Gene Name
sex comb on midleg homolog 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031694.2 Intron NP_001026864.1
NM_001172218.1 Intron NP_001165689.1
NM_001172219.1 Intron NP_001165690.1
NM_001172220.1 Intron NP_001165691.1
NM_001172221.1 Intron NP_001165692.1
NM_001172222.2 Intron NP_001165693.1
NM_012236.3 Intron NP_036368.1
XM_006710462.2 Intron XP_006710525.1
XM_006710464.1 Intron XP_006710527.1
XM_011541032.2 Intron XP_011539334.1
XM_011541033.2 Intron XP_011539335.1
XM_011541034.1 Intron XP_011539336.1
XM_011541035.2 Intron XP_011539337.1
XM_011541036.2 Intron XP_011539338.1
XM_011541039.1 Intron XP_011539341.1
XM_011541040.2 Intron XP_011539342.1
XM_011541043.2 Intron XP_011539345.1
XM_011541044.1 Intron XP_011539346.1
XM_011541045.2 Intron XP_011539347.2
XM_017000698.1 Intron XP_016856187.1
XM_017000699.1 Intron XP_016856188.1
XM_017000700.1 Intron XP_016856189.1
XM_017000701.1 Intron XP_016856190.1
XM_017000702.1 Intron XP_016856191.1
XM_017000703.1 Intron XP_016856192.1
XM_017000704.1 Intron XP_016856193.1
XM_017000705.1 Intron XP_016856194.1
XM_017000706.1 Intron XP_016856195.1
XM_017000707.1 Intron XP_016856196.1
XM_017000708.1 Intron XP_016856197.1
XM_017000709.1 Intron XP_016856198.1
XM_017000710.1 Intron XP_016856199.1
XM_017000711.1 Intron XP_016856200.1
XM_017000712.1 Intron XP_016856201.1
XM_017000713.1 Intron XP_016856202.1
Gene
SLFNL1
Gene Name
schlafen like 1
There are no transcripts associated with this gene.

Gene
SLFNL1-AS1
Gene Name
SLFNL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details