Product Details
- SNP ID
-
rs11643696
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:54926407 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTTTAATATTAAAGAAAAACATA[C/T]ACACTTCTTACATACACACTGATAG
- Phenotype
-
MIM: 615624
MIM: 606195
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRNDE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111640757] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRNDE
- Gene Name
- colorectal neoplasia differentially expressed (non-protein coding)
- Gene
- IRX5
- Gene Name
- iroquois homeobox 5
There are no transcripts associated with this gene.
View Full Product Details