Product Details

SNP ID
rs11812116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5525534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCCGTGTGCTGGTGTCCAAGT[A/G]AGGCCGGCGCCCACCATGCTCCTGG
Phenotype
MIM: 114184
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALML3 PubMed Links

Gene Details

Gene
CALML3
Gene Name
calmodulin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005185.3 646 Silent Mutation TAA,TGA *150* NP_005176.1
Gene
CALML3-AS1
Gene Name
CALML3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details