Product Details

SNP ID
rs11595614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5525521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACTACGAGGAGTTTGTCCGTGTG[A/C]TGGTGTCCAAGTGAGGCCGGCGCCC
Phenotype
MIM: 114184
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALML3 PubMed Links

Gene Details

Gene
CALML3
Gene Name
calmodulin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005185.3 633 Missense Mutation ATG,CTG M146L NP_005176.1
Gene
CALML3-AS1
Gene Name
CALML3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details