Product Details

SNP ID
rs10881895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91178262 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTCTAAAATTAATAAATCAAGA[C/T]GGCAGTTTAAACATTTAAGAATATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCGF5 PubMed Links

Gene Details

Gene
PCGF5
Gene Name
polycomb group ring finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256549.1 Intron NP_001243478.1
NM_001257101.1 Intron NP_001244030.1
NM_032373.4 Intron NP_115749.2
XM_006718030.1 Intron XP_006718093.1
XM_011540271.2 Intron XP_011538573.1
XM_011540272.2 Intron XP_011538574.1
XM_017016776.1 Intron XP_016872265.1
XM_017016777.1 Intron XP_016872266.1
XM_017016778.1 Intron XP_016872267.1

View Full Product Details