Product Details

SNP ID
rs12721609
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.3:119781836 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AAATGCACTCAGAATTACTTAGACA[A/G]AGCGGATATTTGCCACTCTCTTCCC
Phenotype
MIM: 603065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR1I2 PubMed Links

Gene Details

Gene
NR1I2
Gene Name
nuclear receptor subfamily 1 group I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003889.3 1353 UTR 5 NP_003880.3
NM_022002.2 1353 Intron NP_071285.1
NM_033013.2 1353 UTR 5 NP_148934.1

View Full Product Details