Product Details

SNP ID
rs11964212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87408363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGTTTCGCTCCACTGGCTACC[C/G]CAGAAGCCGGACCTCGGGAGACTTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP206 PubMed Links

Gene Details

Gene
CFAP206
Gene Name
cilia and flagella associated protein 206
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031743.2 Intron NP_001026913.1
Gene
LINC01590
Gene Name
long intergenic non-protein coding RNA 1590
There are no transcripts associated with this gene.

View Full Product Details