Product Details
- SNP ID
-
rs12193964
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:89833643 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCCCTGTGTTGTTTTTTTTTTTT[C/T]CGATTTATAAATAGTTTATCCTGTT
- Phenotype
-
MIM: 606880
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CASP8AP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60722861] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASP8AP2
- Gene Name
- caspase 8 associated protein 2
- Gene
- MDN1
- Gene Name
- midasin AAA ATPase 1
There are no transcripts associated with this gene.
View Full Product Details