Product Details

SNP ID
rs13140786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:26589364 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCATAGATCTTAGGGTACTACAG[A/G]GTTTTTCTCATTGTTTCTACTACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D19 PubMed Links

Gene Details

Gene
TBC1D19
Gene Name
TBC1 domain family member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292054.1 Intron NP_001278983.1
NM_018317.3 Intron NP_060787.2
XM_011513852.2 Intron XP_011512154.1
XM_017008363.1 Intron XP_016863852.1
XM_017008364.1 Intron XP_016863853.1

View Full Product Details