Product Details

SNP ID
rs13273942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73997207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCATCTCATCATCCACCTAATTC[A/C]TTAGGCCCCAAACCAGAAGTATCCT
Phenotype
MIM: 605243
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LY96 PubMed Links

Gene Details

Gene
LY96
Gene Name
lymphocyte antigen 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195797.1 Intron NP_001182726.1
NM_015364.4 Intron NP_056179.3
XM_011517508.2 Intron XP_011515810.1
XM_017013299.1 Intron XP_016868788.1
XM_017013300.1 Intron XP_016868789.1

View Full Product Details