Product Details

SNP ID
rs11572321
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:59896720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCATCTCTATTGTGTGTATTTTT[A/T]GTAGCTTACTTTCTCCCTAAGTAGT
Phenotype
MIM: 601258
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP2J2 PubMed Links

Gene Details

Gene
CYP2J2
Gene Name
cytochrome P450 family 2 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000775.3 Intron NP_000766.2

View Full Product Details