Product Details

SNP ID
rs11186740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91908383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTTCCTCTCTGAGGGGTTTTCT[C/T]TGGGCGGTGCGCTTTGGGGAGGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFBP3 PubMed Links

Gene Details

Gene
FGFBP3
Gene Name
fibroblast growth factor binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152429.4 769 Missense Mutation AAA,AGA K196R NP_689642.3

View Full Product Details