Product Details

SNP ID
rs10881999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91927514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATATTTATATTTTGTAATTGGTT[A/G]ATTGTTCTCTTACTTGCTGGATTAA
Phenotype
MIM: 605191
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTAF1 PubMed Links

Gene Details

Gene
BTAF1
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003972.2 Intron NP_003963.1
XM_011540326.1 Intron XP_011538628.1
XM_011540327.2 Intron XP_011538629.1
XM_011540328.2 Intron XP_011538630.1
XM_011540329.2 Intron XP_011538631.1
XM_017016877.1 Intron XP_016872366.1
XM_017016878.1 Intron XP_016872367.1
XM_017016879.1 Intron XP_016872368.1
XM_017016880.1 Intron XP_016872369.1
XM_017016881.1 Intron XP_016872370.1

View Full Product Details