Product Details
- SNP ID
-
rs11202539
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:87756473 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAAATACATTTTCGTTAACAAAT[C/T]GAGGTCTAGTCTTCAATATCTCCAA
- Phenotype
-
MIM: 614452
MIM: 603005
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATAD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115112112] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATAD1
- Gene Name
- ATPase family, AAA domain containing 1
- Gene
- PAPSS2
- Gene Name
- 3'-phosphoadenosine 5'-phosphosulfate synthase 2
There are no transcripts associated with this gene.
View Full Product Details