Product Details

SNP ID
rs11202863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88820548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGACAACCTCCTGAGGGATGGGG[C/T]TAGGACCCATGAAGGCAGAATTACG
Phenotype
MIM: 613923
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD22 PubMed Links

Gene Details

Gene
ANKRD22
Gene Name
ankyrin repeat domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144590.2 3337 UTR 3 NP_653191.2
Gene
LIPM
Gene Name
lipase family member M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128215.1 3337 Intron NP_001121687.1
XM_011539748.2 3337 UTR 3 XP_011538050.1
XM_011539751.2 3337 UTR 3 XP_011538053.1
XM_011539752.2 3337 UTR 3 XP_011538054.1
XM_011539753.2 3337 Intron XP_011538055.1

View Full Product Details