Product Details

SNP ID
rs13152253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122702989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTAAAAATTTTGTTCCACAATT[A/G]TTATAGTAAAGTATAAAACTTTTAT
Phenotype
MIM: 610683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BBS12 PubMed Links

Gene Details

Gene
BBS12
Gene Name
Bardet-Biedl syndrome 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178007.1 Intron NP_001171478.1
NM_152618.2 Intron NP_689831.2
XM_011531680.2 Intron XP_011529982.1
XM_017007831.1 Intron XP_016863320.1

View Full Product Details