Product Details

SNP ID
rs13389423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:214728827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTGTGTGCAGAAGCGCTGATCA[A/G]AATCGGGTCTCGCATGGTATGCGAC
Phenotype
MIM: 601593
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BARD1 PubMed Links

Gene Details

Gene
BARD1
Gene Name
BRCA1 associated RING domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000465.3 786 Missense Mutation TCT,TTT S728F NP_000456.2
NM_001282543.1 786 Missense Mutation TCT,TTT S709F NP_001269472.1
NM_001282545.1 786 Missense Mutation TCT,TTT S277F NP_001269474.1
NM_001282548.1 786 Missense Mutation TCT,TTT S258F NP_001269477.1
NM_001282549.1 786 Missense Mutation TCT,TTT S215F NP_001269478.1
XM_017004613.1 786 Missense Mutation TCT,TTT S761F XP_016860102.1
XM_017004614.1 786 Intron XP_016860103.1

View Full Product Details