Product Details

SNP ID
rs13012441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197738397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGCCTAAAACTTGATTCTGTAT[A/G]AGATGTTACTGCTGAATTAGGGAAT
Phenotype
MIM: 606165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BOLL PubMed Links

Gene Details

Gene
BOLL
Gene Name
boule homolog, RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284358.1 Intron NP_001271287.1
NM_001284361.1 Intron NP_001271290.1
NM_001284362.1 Intron NP_001271291.1
NM_033030.5 Intron NP_149019.1
NM_197970.2 Intron NP_932074.1
XM_006712715.3 Intron XP_006712778.1
XM_011511692.2 Intron XP_011509994.1
XM_011511693.2 Intron XP_011509995.1
XM_011511694.2 Intron XP_011509996.1
XM_017004773.1 Intron XP_016860262.1

View Full Product Details