Product Details
- SNP ID
-
rs13250483
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:95248498 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTTTCTGATGAATTAACTATAGAA[A/T]ATTTTCATTAGCTTCAATGATCTGT
- Phenotype
-
MIM: 614477
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C8orf37
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80034424] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C8orf37
- Gene Name
- chromosome 8 open reading frame 37
View Full Product Details