Product Details

SNP ID
rs9331944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27597514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCAACAGCTTTTACAAATAAAA[C/T]CGCTGCAAAAGCAATAGCTCTTACA
Phenotype
MIM: 185430 MIM: 132811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLU PubMed Links

Gene Details

Gene
CLU
Gene Name
clusterin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001831.3 2263 UTR 3 NP_001822.3
XM_006716284.2 2263 Intron XP_006716347.1
Gene
EPHX2
Gene Name
epoxide hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256482.1 2263 Intron NP_001243411.1
NM_001256483.1 2263 Intron NP_001243412.1
NM_001256484.1 2263 Intron NP_001243413.1
NM_001979.5 2263 Intron NP_001970.2
XM_017013199.1 2263 Intron XP_016868688.1
XM_017013200.1 2263 Intron XP_016868689.1

View Full Product Details