Product Details

SNP ID
rs11891467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25927215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCAAGTGATGAATGAGGTGATA[C/T]GTGCTGCTTTGTCCACAAACGAGAA
Phenotype
MIM: 602845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF3C PubMed Links

Gene Details

Gene
KIF3C
Gene Name
kinesin family member 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002254.6 4799 UTR 3 NP_002245.4
XM_005264299.2 4799 UTR 3 XP_005264356.2

View Full Product Details