Product Details

SNP ID
rs12055741
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108297931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTGAATAGCCCATAGTCTTGA[A/G]AGGCATATGGAATTATGGTAGCCAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LACE1 PubMed Links

Gene Details

Gene
LACE1
Gene Name
lactation elevated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323005.1 Intron NP_001309934.1
NM_145315.4 Intron NP_660358.2
XM_005266885.3 Intron XP_005266942.1
XM_011535657.2 Intron XP_011533959.1
XM_011535659.2 Intron XP_011533961.1
XM_011535660.2 Intron XP_011533962.1
XM_011535661.2 Intron XP_011533963.1
XM_017010647.1 Intron XP_016866136.1
XM_017010648.1 Intron XP_016866137.1
XM_017010649.1 Intron XP_016866138.1

View Full Product Details