Product Details

SNP ID
rs6838948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56914436 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGTAAAAGAATTTGTAGTCCCT[G/T]TGGAATTTGTCATAAGCTGTGATTG
Phenotype
MIM: 600571
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
REST PubMed Links

Gene Details

Gene
REST
Gene Name
RE1 silencing transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193508.1 Intron NP_001180437.1
NM_005612.4 Intron NP_005603.3
XM_011534401.2 Intron XP_011532703.1
XM_017008527.1 Intron XP_016864016.1

View Full Product Details