Product Details

SNP ID
rs11109512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98523010 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATATCATAGAGCTGTTGGGTTTT[A/G]AAGTTGAAAAATACGTTAGAGATCA
Phenotype
MIM: 188380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPO PubMed Links

Gene Details

Gene
TMPO
Gene Name
thymopoietin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032283.2 Intron NP_001027454.1
NM_001032284.2 Intron NP_001027455.1
NM_001307975.1 Intron NP_001294904.1
NM_003276.2 Intron NP_003267.1
XM_005269132.3 Intron XP_005269189.1
XM_017019914.1 Intron XP_016875403.1
Gene
TMPO-AS1
Gene Name
TMPO antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details