Product Details

SNP ID
rs13019825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200594015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGAGTTTCTTTCTTTCTCATTG[G/T]GTTAGTTCAGGTTCTTCAGGATGTG
Phenotype
MIM: 602841 MIM: 612425
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AOX1 PubMed Links

Gene Details

Gene
AOX1
Gene Name
aldehyde oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159.3 Intron NP_001150.3
XM_011511062.1 Intron XP_011509364.1
XM_017003946.1 Intron XP_016859435.1
XM_017003947.1 Intron XP_016859436.1
Gene
SGO2
Gene Name
shugoshin 2
There are no transcripts associated with this gene.

View Full Product Details