Product Details

SNP ID
rs12133516
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:178748517 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAACTATTTATTGATAGTCACA[A/G]CAACAGGGATGAGGTCTCAAAATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RALGPS2 PubMed Links

Gene Details

Gene
RALGPS2
Gene Name
Ral GEF with PH domain and SH3 binding motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286247.1 Intron NP_001273176.1
NM_152663.4 Intron NP_689876.2
XM_006711410.3 Intron XP_006711473.1
XM_006711411.3 Intron XP_006711474.1
XM_011509688.1 Intron XP_011507990.1
XM_017001591.1 Intron XP_016857080.1

View Full Product Details