Product Details

SNP ID
rs12426224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:66351858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCGATGCCGTATCTGTCTCCTA[A/G]AGTCGCTGGTTTAATTGGTCTGGCA
Phenotype
MIM: 604597 MIM: 614539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIP1 PubMed Links

Gene Details

Gene
GRIP1
Gene Name
glutamate receptor interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178074.1 Intron NP_001171545.1
NM_021150.3 Intron NP_066973.2
XM_005268754.4 Intron XP_005268811.1
XM_005268757.4 Intron XP_005268814.1
XM_011538093.2 Intron XP_011536395.1
XM_017019098.1 Intron XP_016874587.1
XM_017019099.1 Intron XP_016874588.1
XM_017019100.1 Intron XP_016874589.1
XM_017019101.1 Intron XP_016874590.1
XM_017019102.1 Intron XP_016874591.1
Gene
HELB
Gene Name
DNA helicase B
There are no transcripts associated with this gene.

View Full Product Details