Product Details

SNP ID
rs6784630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57233540 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCAGATTTTTTTTAAGAATGGCA[C/G]TAGCCAAACAAGATTTCCTGATGCT
Phenotype
MIM: 604299 MIM: 601802
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012096.2 Intron NP_036228.1
XM_011533583.2 Intron XP_011531885.1
Gene
HESX1
Gene Name
HESX homeobox 1
There are no transcripts associated with this gene.

View Full Product Details