Product Details

SNP ID
rs10797787
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:182600349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACAGAGTGGCAGAGGCGGCTGA[A/G]GCTTGGGCAGCCAGGTCCCGGTAAG
Phenotype
MIM: 602514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS16 PubMed Links

Gene Details

Gene
RGS16
Gene Name
regulator of G-protein signaling 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002928.3 706 Silent Mutation GCC,GCT A184A NP_002919.3

View Full Product Details