Product Details

SNP ID
rs12565393
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:183259171 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGCAGATTCCTAAGCCCACCCC[C/T]CATATATTCTGACTCAGTGGGACTG
Phenotype
MIM: 608701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NMNAT2 PubMed Links

Gene Details

Gene
NMNAT2
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015039.3 Intron NP_055854.1
NM_170706.3 Intron NP_733820.1

View Full Product Details