Product Details

SNP ID
rs12907488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:36894147 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCTGTCTTTCTGTCTCATGTCG[A/G]AAGTCAGTAGGCACATGTAAAAAAT
Phenotype
MIM: 601740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC145845 PubMed Links

Gene Details

Gene
LOC145845
Gene Name
uncharacterized LOC145845
There are no transcripts associated with this gene.

Gene
MEIS2
Gene Name
Meis homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220482.1 Intron NP_001207411.1
NM_002399.3 Intron NP_002390.1
NM_170674.4 Intron NP_733774.1
NM_170675.4 Intron NP_733775.1
NM_170676.4 Intron NP_733776.1
NM_170677.4 Intron NP_733777.1
NM_172315.2 Intron NP_758526.1
NM_172316.2 Intron NP_758527.1
XM_006720523.1 Intron XP_006720586.1
XM_006720524.1 Intron XP_006720587.1
XM_006720525.1 Intron XP_006720588.1
XM_006720526.2 Intron XP_006720589.1
XM_006720527.2 Intron XP_006720590.1
XM_006720529.2 Intron XP_006720592.1
XM_011521591.2 Intron XP_011519893.1
XM_017022205.1 Intron XP_016877694.1

View Full Product Details