Product Details

SNP ID
rs12498747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183039061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGGACAGGTATCCACTACCAA[C/T]TAAGAAAGGATCAGCAAGCAGATGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM92A1P2 PubMed Links

Gene Details

Gene
FAM92A1P2
Gene Name
family with sequence similarity 92, member A3
There are no transcripts associated with this gene.

View Full Product Details