Product Details
- SNP ID
-
rs12220909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:14383222 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTTCAGTCAGGCTGTTCCTGTCA[C/G]GCTGGTGTGAAATGAATTAGCAATG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4293
PubMed Links
Gene Details
- Gene
- MIR4293
- Gene Name
- microRNA 4293
There are no transcripts associated with this gene.
View Full Product Details