Product Details

SNP ID
rs10888402
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:151052212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCATAACTAGAGTGGGCCAGGG[G/T]TGCTGCTAAATATCCTGTAATGCAC
Phenotype
MIM: 611275 MIM: 604684
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
There are no transcripts associated with this gene.

Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017860.3 1806 Intron NP_060330.2
Gene
CDC42SE1
Gene Name
CDC42 small effector 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038707.1 1806 UTR 3 NP_001033796.1
NM_020239.3 1806 UTR 3 NP_064624.1
XM_017001847.1 1806 UTR 3 XP_016857336.1
Gene
MLLT11
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
There are no transcripts associated with this gene.

View Full Product Details