Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_024852.3 | Intron | NP_079128.2 | |||
NM_177422.2 | Intron | NP_803171.1 | |||
XM_005270575.3 | Intron | XP_005270632.1 | |||
XM_005270576.3 | Intron | XP_005270633.1 | |||
XM_011540879.2 | Intron | XP_011539181.1 | |||
XM_011540882.2 | Intron | XP_011539184.1 | |||
XM_017000523.1 | Intron | XP_016856012.1 | |||
XM_017000524.1 | Intron | XP_016856013.1 | |||
XM_017000525.1 | Intron | XP_016856014.1 | |||
XM_017000526.1 | Intron | XP_016856015.1 | |||
XM_017000527.1 | Intron | XP_016856016.1 | |||
XM_017000528.1 | Intron | XP_016856017.1 |