Product Details

SNP ID
rs3218232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101773571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTTTTGGAAAGCTTAACATTTT[C/T]AAACCTATTCTTCCACCCCAGTGAC
Phenotype
MIM: 600483 MIM: 606456
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF8 PubMed Links

Gene Details

Gene
FGF8
Gene Name
fibroblast growth factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206389.1 Intron NP_001193318.1
NM_006119.4 Intron NP_006110.1
NM_033163.3 Intron NP_149353.1
NM_033164.3 Intron NP_149354.1
NM_033165.3 Intron NP_149355.1
Gene
NPM3
Gene Name
nucleophosmin/nucleoplasmin 3
There are no transcripts associated with this gene.

View Full Product Details